Anti-GPR83 Rabbit Polyclonal Antibody
Catalog # AVIVARP59122_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor 83
- Antigen symbol:GPR83
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:10888
- Antigen synonyms:GIR|GPR72
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: GPR83 (G protein-coupled receptor 83)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse