Anti-APOL1 Rabbit Polyclonal Antibody
AVIVARP58856_P050
- Antibody type:Primary
- Antigen name:Apolipoprotein L1
- Antigen symbol:APOL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:8542
- Antigen synonyms:APOL|APO-L|FSGS4|APOL-I
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: APOL1 (apolipoprotein L, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human