Anti-TRIM25 Rabbit Polyclonal Antibody
AVIVARP58105_P050
- Antibody type:Primary
- Antigen name:tripartite motif containing 25
- Antigen symbol:TRIM25
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:7706
- Antigen synonyms:Z147|ZNF147|EFP|RNF147
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: TRIM25 (tripartite motif containing 25)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: