Anti-RAD1 Rabbit Polyclonal Antibody
Catalog # AVIVARP57717_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:RAD1 checkpoint DNA exonuclease
- Antigen symbol:RAD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:5810
- Antigen synonyms:REC1|HRAD1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: RAD1 (RAD1 homolog (S. pombe))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat