Anti-PGM3 Rabbit Polyclonal Antibody
Catalog # AVIVARP56755_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:phosphoglucomutase 3
- Antigen symbol:PGM3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:5238
- Antigen synonyms:IMD23|AGM1|PGM 3|PAGM
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: PGM3 (phosphoglucomutase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: