Anti-HADH Rabbit Polyclonal Antibody
AVIVARP54765_P050
- Antibody type:Primary
- Antigen name:Hydroxyacyl-CoA dehydrogenase, mitochondrial
- Antigen symbol:HADH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:3033
- Antigen synonyms:HADH1|HAD|HADHSC|SCHAD|MSCHAD|HHF4|HCDH
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: HADH (hydroxyacyl-CoA dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: