Anti-ENSA Rabbit Polyclonal Antibody
AVIVARP53514_P050
- Antibody type:Primary
- Antigen name:endosulfine alpha
- Antigen symbol:ENSA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:2029
- Antigen synonyms:ARPP-19e
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ENSA (endosulfine alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: