Anti-ERI2 Rabbit Polyclonal Antibody
AVIVARP52509_P050
- Antibody type:Primary
- Antigen name:ERI1 exoribonuclease family member 2
- Antigen symbol:ERI2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:112479
- Antigen synonyms:EXOD1|ZGRF5
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ERI2 (ERI1 exoribonuclease family member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: