Anti-FUBP3 Rabbit Polyclonal Antibody
Catalog # AVIVARP51747_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Far upstream element binding protein 3
- Antigen symbol:FUBP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:8939
- Antigen synonyms:FBP3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: FUBP3 (far upstream element (FUSE) binding protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: