Anti-GALNT18 Rabbit Polyclonal Antibody
AVIVARP49185_P050
- Antibody type:Primary
- Antigen name:Polypeptide N-Acetylgalactosaminyltransferase 18
- Antigen symbol:GALNT18
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:374378
- Antigen synonyms:GALNT15|GalNAc-T18|GalNAc-T15|GALNTL4|GALNACT18
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: GALNT18 (UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: