Anti-SULT1E1 Rabbit Polyclonal Antibody
Catalog # AVIVARP48669_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:sulfotransferase family 1E, estrogen-preferring, member 1
- Antigen symbol:Sulfotransferase Family 1E Estrogen-Preferring Member 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:6783
- Antigen synonyms:EST|ST1E1|EST-1|STE
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SULT1E1 (sulfotransferase family 1E, estrogen-preferring, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: