Anti-SULT2B1 Rabbit Polyclonal Antibody
AVIVARP48635_P050
- Antibody type:Primary
- Antigen name:sulfotransferase family, cytosolic, 2B, member 1
- Antigen symbol:SULT2B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:6820
- Antigen synonyms:HSST2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: SULT2B1 (sulfotransferase family, cytosolic, 2B, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: