Anti-PRDX1 Rabbit Polyclonal Antibody
Catalog # AVIVARP48454_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:peroxiredoxin 1
- Antigen symbol:PRDX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:5052
- Antigen synonyms:MSP23|PAGA|PAG|NKEF-A|PRXI|NKEFA|PRX1|TDPX2|PAGB
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: PRDX1 (peroxiredoxin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: