Anti-VIMP Rabbit Polyclonal Antibody
Catalog # AVIVARP48082_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:VCP-interacting membrane protein
- Antigen symbol:VIMP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:55829
- Antigen synonyms:SELS|AD-015|SEPS1|ADO15|SBBI8
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: VIMP (VCP-interacting membrane protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: