Anti-VARS2 Rabbit Polyclonal Antibody
Catalog # AVIVARP47652_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:valyl-tRNA synthetase 2, mitochondrial
- Antigen symbol:VARS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:57176
- Antigen synonyms:COXPD20|VALRS|VARS2L|VARSL
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: VARS2 (valyl-tRNA synthetase 2, mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: