Anti-PEMT Rabbit Polyclonal Antibody
Catalog # AVIVARP43558_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:phosphatidylethanolamine N-methyltransferase
- Antigen symbol:PEMT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:10400
- Antigen synonyms:PNMT|PEMPT|PEAMT|PEMT2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: PEMT (phosphatidylethanolamine N-methyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: