Anti-PKLR Rabbit Polyclonal Antibody
AVIVARP41699_T100
- Antibody type:Primary
- Antigen name:pyruvate kinase, liver and RBC
- Antigen symbol:PKLR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:5313
- Antigen synonyms:PKL|RPK|PKRL|PKR|PK1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: PKLR (pyruvate kinase, liver and RBC)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: