Anti-EIF2A Rabbit Polyclonal Antibody
Catalog # AVIVARP41041_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Eukaryotic translation initiation factor 2A, 65kDa
- Antigen symbol:EIF2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:83939
- Antigen synonyms:MST089|EIF-2A|MSTP089|MSTP004|CDA02
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: EIF2A (eukaryotic translation initiation factor 2A, 65kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: