Anti-LARP7 Rabbit Polyclonal Antibody
Catalog # AVIVARP40848_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:La ribonucleoprotein domain family, member 7
- Antigen symbol:LARP7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:51574
- Antigen synonyms:HDCMA18P|ALAZS|PIP7S
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: LARP7 (La ribonucleoprotein domain family, member 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: