Anti-POP4 Rabbit Polyclonal Antibody
Catalog # AVIVARP40674_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:POP4 homolog, ribonuclease P/MRP subunit
- Antigen symbol:POP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:10775
- Antigen synonyms:RPP29
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: POP4 (processing of precursor 4, ribonuclease P/MRP subunit (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: