Anti-ZNF296 Rabbit Polyclonal Antibody
AVIVARP39858_P050
- Antibody type:Primary
- Antigen name:zinc finger protein 296
- Antigen symbol:ZNF296
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:162979
- Antigen synonyms:ZFP296|ZNF342
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ZNF296 (zinc finger protein 296)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human