Anti-MSL3 Rabbit Polyclonal Antibody
Catalog # AVIVARP39783_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:male-specific lethal 3 homolog (Drosophila)
- Antigen symbol:MSL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:10943
- Antigen synonyms:MSL3L1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: MSL3 (male-specific lethal 3 homolog (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat