Anti-TAF7L Rabbit Polyclonal Antibody
AVIVARP37402_T100
- Antibody type:Primary
- Antigen name:TAF7-like RNA polymerase II, TATA box binding protein (TBP)-associated factor
- Antigen symbol:TAF7L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:74469
- Antigen synonyms:Taf2q|4933438I11Rik
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: TAF7L (TAF7-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 50kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Mouse