Anti-THRB Rabbit Polyclonal Antibody
Catalog # AVIVARP36994_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Thyroid Hormone Receptor, beta
- Antigen symbol:THRB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- ChIP:Yes
- Gene ID:21834
- Antigen synonyms:T3R[b]|c-erbAbeta|Nr1a2|T3Rbeta|Thrb1|Thrb2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: THRB (thyroid hormone receptor, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat