Anti-FOXA3 Rabbit Polyclonal Antibody
Catalog # AVIVARP36881_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Forkhead Box A3
- Antigen symbol:FOXA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:15377
- Antigen synonyms:Hnf-3g|Tcf3g|Hnf3g|Tcf-3g
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: FOXA3 (forkhead box A3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: