Anti-ZFP42 Rabbit Polyclonal Antibody
Catalog # AVIVARP36245_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:ZFP42 zinc finger protein
- Antigen symbol:ZFP42
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:132625
- Antigen synonyms:zfp-42|REX1|ZNF754|REX-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ZFP42 (ZFP42 zinc finger protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human