Anti-TRPV4 Rabbit Polyclonal Antibody
AVIVARP35416_P050
- Antibody type:Primary
- Antigen name:transient receptor potential cation channel, subfamily V, member 4
- Antigen symbol:TRPV4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:59341
- Antigen synonyms:SMAL|VRL2|SSQTL1|TRP12|VROAC|CMT2C|OTRPC4|HMSN2C|BCYM3|SPSMA
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: TRPV4 (transient receptor potential cation channel, subfamily V, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: