Anti-RACGAP1 Rabbit Polyclonal Antibody
Catalog # AVIVARP31905_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Rac GTPase activating protein 1
- Antigen symbol:RACGAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:29127
- Antigen synonyms:MgcRacGAP|HsCYK-4|ID-GAP|CYK4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: RACGAP1 (Rac GTPase activating protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: