Anti-CHRM1 Rabbit Polyclonal Antibody
BOSSBS-1150R
: Bioss
- Antibody type:Primary
- Antigen name:Cholinergic receptor, Muscarinic 1
- Antigen symbol:CHRM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Antigen synonyms:HM1|M1R|M1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Primary
Antigen: CHRM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: