Anti-Neurogenin 3 Rabbit Polyclonal Antibody
BOSSBS-0922R
: Bioss
- Antibody type:Primary
- Antigen name:Neurogenin 3
- Antigen symbol:NEUROG3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Antigen synonyms:NGN|bHLHa7|Atoh5|NGN1|Neurogenic basic-helix-loop-helix- protein|NGN-3|ngn3|Neurogenic differentiation factor 3|NEUROG1|NeuroD3|NEUROD3|Math4B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: Neurogenin 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat