Order Entry
United Kingdom
ContactUsLinkComponent
Ampoule snapper, AccuVac®
Catalog # HACH2405200
Supplier:  Hach
undefined
Ampoule snapper, AccuVac®
Catalog # HACH2405200
Supplier:  Hach
Specifications
  • Description:
    Snapper for ampoules
  • Seq:
    01
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
This snapper is used for opening ampoules immersed in samples.
  • Avoids sample contact
  • Used with colorimeters and spectrophotometers