Bottle Carrier
Catalog # SAFUWBC80T
Supplier: SAFETY UNLIMITED
Specifications
- Description:Bottle carrier, E-WBC80T
- W×D×H:635×332×381 mm
- Environmentally Preferable:
- Pk:1
Specifications
About this item
Zintec coated steel.
- Carrying handles
- Castors for mobility
- Hasp and staple for locking
These bottle carriers are suitable for storing, protecting and transporting winchester bottles with epoxy powder coating.