Genomic DNA isolation and purification kits, GET™ Plant DNA Template
Catalog # 786-355
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:100 Tests
- Application:
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Total DNA
- Sample type:Plant samples
- Target molecule:DNA
- Description:GET™ Plant DNA Template
- Environmentally Preferable:
- Description:GET™ Plant DNA Template
Specifications
About this item
GET™ Plant DNA Template is a spin column format kit suitable for the preparation of DNA templates from plant samples. The isolated genomic DNA is suitable for PCR and other applications. GET™ Plant DNA Template purifies plant genomic DNA free from plant impurities known to inhibit downstream applications. The high yield, extracted DNA is normally >50 kb in size. The yield is ~30 µg DNA per preparation, depending on the plant source and quantity used.