Disposal bins for serological pipettes, Bio-bin®
The 6 L Bio-bin® has been specifically designed to safely contain serological pipettes in labs.
- Tall and thin: Best used with a stand
- Fold-flat space efficient design, stackable pre- and post-use
- Absorbent Bio-matt™ in the base of every bin can absorb 100 ml of fluid
- Built-in lid and handles
- Temporary and permanent seals
- Fully autoclavable
Frequently Bought Together