Genomic DNA extraction kits, plant gDNA kit, Axygen® AxyPrep™ Mag
AXYGMAGPLANTGDNA10
: Corning
:
- Pk:1
- Application:
- Method / Format:Magnetic beads
- Prep size:Mini
- Refine target molecule:Total DNA
- Sample type:Plant samples
- Target molecule:DNA
- Description:Axygen® AxyPrep™ Mag Plant gDNA Kit, 10 ml
- Seq:01
- Environmentally Preferable:
- Description:Axygen® AxyPrep™ Mag Plant gDNA Kit
Axygen® AxyPrep™ Mag Plant gDNA Kit is a magnetic bead-based purification system that allows rapid and reliable isolation of high quality gDNA from a wide variety of plant samples. Up to 96 samples (50 mg fresh tissue per sample or 15 mg of dried tissue per sample) can be processed in less than one hour. The Axygen® AxyPrep™ Mag Plant gDNA Kit eliminates polysaccharides, phenolic compounds and enzyme inhibitors with no organic extractions. This kit is adaptable for use on major liquid handling workstations for high throughput processing.
- High quality, high yield DNA
- Non alcohol based purification
- Compatible with genomic downstream applications: PCR, Sanger and next generation sequencing, fragment analysis, genotyping and SNP detection, and restriction enzyme clean up