Bulb pipettes, MBL®, two marks, Class AS
Supplier: SciLabware
Certificates
About this item
Soda lime glass.
- Calibrated to deliver (TD, Ex)
- Permanent amber stain marking
- With +5 s waiting time
- Features a safety bulb at the top of the pipette tube
- Printed with batch code for downloadable batch certificate
Certifications: Comply with ISO 648 and DIN 12690
Frequently Bought Together