TLC plates, silica gel 60, unmodified layers, aluminium sheets, ALUGRAM® Xtra SILG
Supplier: MACHEREY-NAGEL
Certificates
About this item
The highly polymeric binder used for these sheets is stable in almost all organic solvents and resistant towards aggressive visualisation reagents. It is also completely stable in purely aqueous eluents.
- Precise colourisation results, even with 100% aqueous detection reagents
- Easy and reliable cutting due to an optimised binder system, no flaking of silica
Silica 60: specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 5 to 17 μm.