TruCLEAN Pro® cleaning systems work to capture and isolate contaminants, ensuring the delivery of unadulterated cleaning and sanitizing agents. TruCLEAN components are constructed of high-grade stainless steel, are easy to maintain, and guaranteed to deliver reliable cleaning results time after time.
- Reduce the risk of cross-contamination
- 100% compatible with gamma, ETO autoclave sterilisation
- Avoid run-off contaminants, common with steam-sterilisers
TruCLEAN Pro® mops:
Designed specifically for use in a cleanroom or sterile environment, TruCLEAN Pro® sponge mops and microfiber mops are ideal for applying disinfectants and sterilants. In today’s cleaning industry the most environmentally friendly, non-chemical tool on the market is microfiber. Autoclave or launder for cleaning. Mop frames are made of high-grade ANSI stainless steel.
Specially formulated polyurethane foam has excellent chemical and microbial resistance
Microfiber mops can absorb up to seven times their weight in liquids
Low particle generation; excellent surface coherence
Can be reused up to 500 times
Quick-release frame-locking mechanism allows easy mop head replacement
Mop frames are made of high-grade AISI stainless steel
All components are compatible with gamma, ETO, and autoclave sterilisation
Designed specifically for use in a cleanroom or sterile environment, TruCLEAN Pro® sponge mops and microfiber mops are ideal for applying disinfectants and sterilants. In today’s cleaning industry the most environmentally friendly, non-chemical tool on the market is microfiber. Autoclave or launder for cleaning. Mop frames are made of high-grade ANSI stainless steel.