Order Entry
United Kingdom
ContactUsLinkComponent
Centrifuge tubes, ASTM, cylindrical, conical bottom
Centrifuge tubes, ASTM, cylindrical, conical bottom
Catalog # HECH40930010
undefined
Centrifuge tubes, ASTM, cylindrical, conical bottom
Catalog # HECH40930010
Specifications
  • Closure type:
    Without cap
  • Capacity:
    100 ml
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Borosilicate glass 3.3, graduated.
  • Withstands forces up to 700×g
  • Design, accuracy, etc. to meet ASTM D 96 and DIN 51793 specifications