Reversed phase HPLC columns, Atlantis® T3
: WATERS
Atlantis T3 columns are universal, silica based, reversed phase C18 columns that provide balanced retention of polar and hydrophobic molecules and useful for developing separations of polar and non-polar compounds.
Frequently Bought Together