Laboratory trays, HDPE
: SEMADENI
Laboratory trays made of white HDPE.
- Stackable
- Good chemical resistance
- Good temperature resistance
With reinforcing ribs and supporting edge.
Frequently Bought Together
Laboratory trays made of white HDPE.
With reinforcing ribs and supporting edge.