Order Entry
United Kingdom
ContactUsLinkComponent
Surgical gloves, latex, sterile, Biogel® Surgeons
Surgical gloves, latex, sterile, Biogel® Surgeons
Certificates
undefined
Surgical gloves, latex, sterile, Biogel® Surgeons
Specifications
  • Catalog No:
  • 112-3330
  • 112-3324
  • 112-3328
  • 112-3326
  • 112-3325
  • 112-3333
  • 112-3332
  • 112-3335
  • 112-3334
  • 112-3327
  • 112-3331
  • Model:
  • Surgeons
  • Surgeons
  • Surgeons
  • Surgeons
  • Surgeons
  • Surgeons
  • Surgeons
  • Surgeons
  • Surgeons
  • Surgeons
  • Surgeons
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Micro-textured
  • Smooth
  • Smooth
  • Smooth
  • Smooth
  • Micro-textured
  • Micro-textured
  • Micro-textured
  • Micro-textured
  • Smooth
  • Micro-textured
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,21 mm
  • 0,21 mm
  • 0,21 mm
  • 0,21 mm
  • 0,21 mm
  • 0,21 mm
  • 0,21 mm
  • 0,21 mm
  • 0,21 mm
  • 0,21 mm
  • 0,21 mm
  • Finger thickness:
  • 0,27 mm
  • 0,27 mm
  • 0,27 mm
  • 0,27 mm
  • 0,27 mm
  • 0,27 mm
  • 0,27 mm
  • 0,27 mm
  • 0,27 mm
  • 0,27 mm
  • 0,27 mm
  • Palm thickness:
  • 0,26 mm
  • 0,26 mm
  • 0,26 mm
  • 0,26 mm
  • 0,26 mm
  • 0,26 mm
  • 0,26 mm
  • 0,26 mm
  • 0,26 mm
  • 0,26 mm
  • 0,26 mm
  • EN norm:
  • EN 455
  • EN 455
  • EN 455
  • EN 455
  • EN 455
  • EN 455
  • EN 455
  • EN 455
  • EN 455
  • EN 455
  • EN 455