Eosin Y (yellowish) 0,5% alcoholic solution Eosin Y-solution 0.5% alcoholic for microscopy, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Solvent Red 43, Field's Stain B, Eosin 225, 2,4,5,7-Tetrabromofluorescein disodium salt, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Tetrabromofluorescein disodium salt, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Eosin Y disodium salt
CAS Number:17372-87-1
:
Formula:C₂₀H₆Br₄Na₂O₅
MDL Number:MFCD00005040
Density:0.8889 g/cm3 (20 °C)
Molecular Weight:691.86 g/mol
Flash Pt:19.1 °C
ADR:3 II
Merck Index:13,03634
:M-Clarity™ quality level = MQ300
UN:1170
:GHS02
:GHS07
Storage Temperature:Ambient
:1
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Flash Point:19.1 °C
- Density:0.8889 g/cm3 (20 °C)
Specification Test Results
Related Information
Frequently Bought Together