Order Entry
United Kingdom
ContactUsLinkComponent
 
Cell counter cups, Coulter®
  DELT200103
 :  DELTALAB
Cell counter cups, Coulter®
  DELT200103
 :  DELTALAB

 

  • Capacity:
    20 ml
  • Description:
    Cell counter cups and lid, Ø: 30 mm, chamber depth: 56 mm
  • Seq:
    02
  • Environmentally Preferable:
  • Pk:
    1.000

 

 

Single use PS cups suitable for cell counting.