Order Entry
United Kingdom
ContactUsLinkComponent
Intramuscular safety needles, BD SafetyGlide™
  BDAM305901
undefined
Intramuscular safety needles, BD SafetyGlide™
  BDAM305901
  • Colour code:
    Orange
  • Gauge:
    25
  • Length:
    15,9 mm
  • Seq:
    01
  • Environmentally Preferable:
  • Pk:
    500
Detachable, disposable, shielding hypodermic needle, stainless steel latch. The BD SafetyGlide™ shielding hypodermic needle is a single-handed, needle-based safety device that can be used with any size conventional syringe. Detachable needle fits on any size conventional syringe, which helps to facilitate standardisation and SKU reduction.
  • It is complemented for Insulin, TB and Allergy for small dose of medication
  • Virtually no splatter upon activation
  • Secure and visible lock