Order Entry
United Kingdom
ContactUsLinkComponent
Recirculating chiller for ICP/ICP-MS
Recirculating chiller for ICP/ICP-MS
  PERKN0772025
Recirculating chiller for ICP/ICP-MS
  PERKN0772025

 

  • Temperature Range:
    −15...40 °C
  • Description:
    Chiller for ICP/ICP-MS, 220-240 V, 50 Hz, 8.5 A
  • Environmentally Preferable:
  • Cooling capacity:
    2,85 kW
  • Pk:
    1
  • Temp. stability:
    ±0,5 °C

 

 

Water cooling systems provide clean, reliable temperature-controlled fluid for open tanks or closed loop cooling and are ideal for many routine process cooling needs.

  • Feature convenient 'one-touch' temperature control and bright LED readouts
  • Instant alerts through audible and visual alarms when operating parameters exceed pre-set limits
  • Built-in safeties to alert low fluid levels and thermal runaway

Other features include a calibration function that allows to adjust the temperature display to match a traceable temperature standard and optional interfaces for various control functions, such as on/off and remote control.