Tough-Spots® laser labels
Supplier: DIVERSIFIED BIOTECH
Certificates
About this item
These Tough-Spots® polyolefin labels are specially top-coated to readily accept laser printing.
- Temperature resistant from −196 to +150 °C
- Chemically inert, resist most organic solvents and caustic agents
- Adhere to most plastics, glass and metals without cracking, peeling or degrading
Tough-Spots® withstand freezer temperatures down to −196 °C (liquid and vapor phase nitrogen), boiling water baths (100 °C), autoclaving and moderate oven temperatures (150 °C).
Frequently Bought Together