Pressure vessels
: Merck
Dispensing pressure vessels hold liquid for filtration through pressure-operated filter holders, if connected to an external pressure source. Used for large-volume filtration, reservoir for buffer or solvent dispensing.
- Made from stainless steel 316
- Closures are secured by cam-lock handle
- Vessels are autoclavable when vented
- Fluoroelastomer gaskets and O-rings used
- Molded neoprene rubber base
Max. operating pressure: 6,9 bar
Max. operating temperature: 121 °C
: Meets ASME®-UM Code requirements.
Frequently Bought Together