Order Entry
United Kingdom
ContactUsLinkComponent
Sodium chloride 1 mol/l (1 N) in aqueous solution
Sodium chloride 1 mol/l (1 N) in aqueous solution
Catalog # PRLS85279.290
Supplier:  VWR Collection
CAS Number:  
Sodium chloride 1 mol/l (1 N) in aqueous solution
Catalog # PRLS85279.290
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Saline, NaCl, Sea salt, Halite
 :
 :58.44 g/mol
Boiling Pt:1413
Melting Pt:801
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477
Merck Index:14,08599