Trimethylaluminium, Sigma-Aldrich®
SIAL663301-25G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
: Trimethylaluminum
:
:72.09 g/mol
:200-853-0
:3394
:75-24-1
:C₃H₉Al
:MFCD00008252
:125 °C (1013 hPa)
:15 °C
:0.748 g/cm³ (20 °C)
:4.2 I
:M-Clarity™ quality level = MQ100